3-Bromo-5-methoxyaniline 98%

Supplier: Ambeed
Warning

A264378-100G A264378-10G A264378-250MG A264378-1G A264378-25G A264378-5G
103812-668EA 10 USD
103812-668 103852-560 76816-054 76816-056 76816-058 77092-214 77092-216 77092-218
3-Bromo-5-methoxyaniline 98%
3-Bromo-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
Flash Pt: 126.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11878497
CAS Number: 16618-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR