3-Bromo-5-iodopyridine 96%

Supplier: Ambeed
Warning

A136702-100G A136702-10G A136702-250MG A136702-1G A136702-25G A136702-5G
103802-334EA 7 USD
103802-334 76996-678 76996-680 76996-682 77315-252 77315-254 77315-256
3-Bromo-5-iodopyridine 96%
3-Bromo-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
MW: 283.89 g/mol
Boiling Pt: 266.5 °C (760 mm Hg)
Melting Pt: 131.3…131.4 °C
Flash Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03086019
CAS Number: 233770-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR