3-Bromo-5-iodophenol 97%

Supplier: Ambeed
Warning

A153696-10G A153696-250MG A153696-25G A153696-1G A153696-5G
103804-554EA 91 USD
103804-554 103842-730 77162-888 77162-890 77162-892 77318-406 77318-408
3-Bromo-5-iodophenol 97%
3-Bromo-5-iodophenol

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₆H₄BrIO
MW: 298.904 g/mol
Boiling Pt: 324.2±32.0 °C (760 mm Hg)
Melting Pt: 82.5 °C
Flash Pt: 149.9 °C
Storage Temperature: Ambient
MDL Number: MFCD12405397
CAS Number: 570391-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR