3-Bromo-5-iodobenzoic acid 97%

Supplier: Ambeed
Warning

A102476-100G A102476-10G A102476-25G A102476-500G A102476-1G A102476-5G
103796-374EA 6 USD
103796-374 103822-068 77107-244 77107-246 77107-248 77107-250 77282-932 77308-950
3-Bromo-5-iodobenzoic acid 97%
3-Bromo-5-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.92 g/mol
Boiling Pt: 385.2 °C (760 mm Hg)
Melting Pt: 218…221 °C
Flash Pt: 186.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00191851
CAS Number: 188815-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR