3-Bromo-5-iodobenzaldehyde 95%

Supplier: Ambeed
Warning

A179901-1G A179901-100MG A179901-250MG
103807-488EA 63 USD
103807-488 103845-842 77083-318 77111-384 77685-407
3-Bromo-5-iodobenzaldehyde 95%
3-Bromo-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrIO
Boiling Pt: 318.433 °C (760 mm Hg)
Flash Pt: 146.384 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779024
CAS Number: 188813-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR