3-Bromo-5-hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A171239-1G A171239-25G A171239-5G A171239-100MG A171239-250MG A171239-10G
103806-608EA 17 USD
103806-608 103844-882 77082-018 77082-020 77082-022 77163-574 77163-576 77892-139
3-Bromo-5-hydroxybenzaldehyde 98%
3-Bromo-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06797981
CAS Number: 199177-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR