3-Bromo-5-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A138625-10G
A138625-1G
A138625-25G
A138625-5G
A138625-100G
103802-640EA
5
USD
103802-640
77272-368
77315-682
77315-684
77315-686
77592-634
3-Bromo-5-fluoropyridine 98%
3-Bromo-5-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₃BrFN Boiling Pt: 152.5 °C (760 mm Hg) Melting Pt: 24…28 °C Flash Pt: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112555 CAS Number: 407-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...