3-Bromo-5-fluoropicolinonitrile 97%

Supplier: Ambeed
Warning

A104980-10G A104980-250MG A104980-25G A104980-1G A104980-5G A104980-100G
103796-808EA 19 USD
103796-808 103822-534 76970-370 76970-372 76970-374 77309-398 77309-400 77880-849
3-Bromo-5-fluoropicolinonitrile 97%
3-Bromo-5-fluoropicolinonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD16988825
CAS Number: 950670-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR