3-Bromo-5-fluoro-2-pyridine carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-carboxy-5-fluoropyridine, 3-Bromo-5-fluoropicolinic acid, 3-Bromo-5-fluoropyridine-2-carboxylic acid

A196172-10G A196172-250MG A196172-1G A196172-25G A196172-5G
103816-822EA 39 USD
103816-822 103847-436 76975-050 76975-052 77085-394 77302-508 77302-510
3-Bromo-5-fluoro-2-pyridine carboxylic acid 97%
3-Bromo-5-fluoro-2-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD12827684
CAS Number: 1189513-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR