3-Bromo-5-fluorophenol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-fluoro-5-hydroxybenzene

A400666-25G A400666-5G A400666-100G A400666-10G A400666-1G A400666-500G
103838-082EA 7 USD
103838-082 76841-536 76841-538 77115-718 77115-720 77115-722 77115-724
3-Bromo-5-fluorophenol 98%
3-Bromo-5-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 231.1 °C (760 mm Hg)
Melting Pt: 36…40 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD07783710
CAS Number: 433939-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR