3-Bromo-5-fluoroisonicotinaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-fluoropyridine-4-carboxaldehyde

A170654-100MG A170654-250MG
103806-522EA 123 USD
103806-522 103844-796 76985-388 76985-390
3-Bromo-5-fluoroisonicotinaldehyde 98%
3-Bromo-5-fluoroisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrFNO
MW: 204 g/mol
Boiling Pt: 227.5±35.0 °C (760 mm Hg)
Flash Pt: 91.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16606512
CAS Number: 1227573-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR