3-Bromo-5-fluorobenzonitrile 98%

Supplier: Ambeed
Danger

A310350-5G A310350-100G A310350-10G A310350-1G A310350-25G
103814-618EA 9 USD
103814-618 103835-800 76838-578 77114-608 77114-610 77306-018 77306-020
3-Bromo-5-fluorobenzonitrile 98%
3-Bromo-5-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrFN
Boiling Pt: 218.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04038227
CAS Number: 179898-34-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR