3-Bromo-5-fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A115003-100G
A115003-10G
A115003-25G
A115003-5G
103824-358EA
9
USD
103824-358
77271-172
77271-174
77271-176
77297-342
3-Bromo-5-fluorobenzoic acid 98%
3-Bromo-5-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrFO₂ Boiling Pt: 303.3 °C (760 mm Hg) Melting Pt: 140 °C Flash Pt: 137.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569540 CAS Number: 176548-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...