3-Bromo-5-fluoroaniline 95%

Supplier: Ambeed
Warning

A100825-100G A100825-10G A100825-250MG
103795-642EA 12 USD
103795-642 103821-338 76765-132 76765-134 76765-136
3-Bromo-5-fluoroaniline 95%
3-Bromo-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 245.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07779529
CAS Number: 134168-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR