Methyl 3-bromo-5-fluoro-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A140050-100MG A140050-250MG
103802-822EA 52 USD
103802-822 77150-026 77150-028
Methyl 3-bromo-5-fluoro-2-methylbenzoate 95%
Methyl 3-bromo-5-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
Boiling Pt: 262.9 °C (760 mm Hg)
Flash Pt: 112.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19440363
CAS Number: 1187318-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR