3-Bromo-5-fluoro-2-methylaniline 98%
Supplier: Ambeed
|
Warning
|
A106869-10G
A106869-25G
A106869-1G
A106869-5G
103797-090EA
11
USD
103797-090
103822-846
77025-494
77025-496
77309-704
77309-706
3-Bromo-5-fluoro-2-methylaniline 98%
3-Bromo-5-fluoro-2-methylaniline
Precaución: for Research Use Only
Formula:
C₇H₇BrFN Boiling Pt: 259 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094193 CAS Number: 502496-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...