3-Bromo-5-fluoro-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A193712-25G A193712-5G A193712-100G A193712-10G
103816-572EA 6 USD
103816-572 103847-186 77085-086 77085-088 77111-892 77111-894
3-Bromo-5-fluoro-2-methoxypyridine 98%
3-Bromo-5-fluoro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
Boiling Pt: 188.3 °C (760 mm Hg)
Flash Pt: 67.715 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972397
CAS Number: 884494-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR