3-Bromo-5-cyanobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A177599-250MG
A177599-100MG
103807-248EA
161
USD
103807-248
103845-578
76974-314
77611-728
3-Bromo-5-cyanobenzoic acid 97%
3-Bromo-5-cyanobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₄BrNO₂ MW: 226.027 g/mol Melting Pt: 182…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD09261037 CAS Number: 453566-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...