3-Bromo-5-chloropyridin-4-ol 95%

Supplier: Ambeed
Warning

A100662-100MG A100662-250MG
103795-558EA 278 USD
103795-558 76646-686 76646-688
3-Bromo-5-chloropyridin-4-ol 95%
3-Bromo-5-chloropyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185351
CAS Number: 1822685-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR