3-Bromo-5-chloroaniline 97%

Supplier: Ambeed
Warning

A228456-1G A228456-25G A228456-5G A228456-100G A228456-10G
103819-398EA 9 USD
103819-398 103850-306 77089-096 77089-098 77089-100 77275-710 77275-712
3-Bromo-5-chloroaniline 97%
3-Bromo-5-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Boiling Pt: 284.1 °C (760 mm Hg)
Melting Pt: 41…45 °C (760 mm Hg)
Flash Pt: 125.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06797804
CAS Number: 96558-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR