3-Bromo-5-chlorophenol 97%

Supplier: Ambeed
Warning

A184930-1G A184930-100G A184930-10G A184930-25G A184930-5G
103815-802EA 8 USD
103815-802 103846-368 77083-998 77274-250 77274-252 77301-932 77301-934
3-Bromo-5-chlorophenol 97%
3-Bromo-5-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Melting Pt: 58 °C
Flash Pt: 110.075 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07780676
CAS Number: 56962-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR