3-Bromo-5-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A125160-1G A125160-5G A125160-25G A125160-10G
103800-636EA 7 USD
103800-636 77312-778 77312-780 77549-480 77666-786
3-Bromo-5-chlorobenzoic acid 98%
3-Bromo-5-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
Boiling Pt: 332 °C (760 mm Hg)
Melting Pt: 190…192 °C
Flash Pt: 154.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06797227
CAS Number: 42860-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR