3-Bromo-5-chlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A123921-100G A123921-10G A123921-25G A123921-1G A123921-5G
103800-464EA 9 USD
103800-464 77108-728 77108-730 77108-732 77108-734 77312-590 77312-592
3-Bromo-5-chlorobenzaldehyde 98%
3-Bromo-5-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO
Boiling Pt: 264 °C (760 mm Hg)
Melting Pt: 68…73 °C
Flash Pt: 113.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06797226
CAS Number: 188813-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR