3-Bromo-5-chloro-2-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A202503-25G A202503-5G A202503-100G A202503-10G
103848-038EA 11 USD
103848-038 77086-142 77086-144 77112-206 77112-208
3-Bromo-5-chloro-2-hydroxybenzaldehyde 97%
3-Bromo-5-chloro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
Boiling Pt: 245.3 °C (760 mm Hg)
Melting Pt: 85…88 °C
Flash Pt: 102.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00051690
CAS Number: 19652-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR