3-Bromo-5-chloro-2-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A128896-10G
A128896-1G
A128896-5G
A128896-100G
103801-198EA
5
USD
103801-198
77109-088
77313-554
77313-556
77549-536
3-Bromo-5-chloro-2-fluoropyridine 98%
3-Bromo-5-chloro-2-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₂BrClFN Boiling Pt: 204 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD04972409 CAS Number: 884494-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...