3-Bromo-5-chloro-1H-pyrrolo[2,3-c]pyridine 96%

Supplier: Ambeed
Warning

A242439-100MG A242439-10G A242439-250MG A242439-1G
103811-468EA 82 USD
103811-468 103851-280 76987-648 76987-650 76987-652 77090-454
3-Bromo-5-chloro-1H-pyrrolo[2,3-c]pyridine 96%
3-Bromo-5-chloro-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405106
CAS Number: 1167056-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR