3-Bromo-5,7-dichloropyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A263519-100MG A263519-250MG
103812-626EA 10 USD
103812-626 103852-514 76722-894 76722-896
3-Bromo-5,7-dichloropyrazolo[1,5-a]pyrimidine 97%
3-Bromo-5,7-dichloropyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂N₃
Melting Pt: 73…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11112133
CAS Number: 114040-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR