3-Bromo-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A230656-25G A230656-5G A230656-100G A230656-10G A230656-250MG A230656-1G
103810-722EA 9 USD
103810-722 103850-464 77089-332 77089-334 77165-506 77165-508 77165-510 77303-882
3-Bromo-5-(trifluoromethyl)pyridine 98%
3-Bromo-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 226.00 g/mol
Boiling Pt: 166 °C (760 mm Hg)
Melting Pt: 44…46 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD04972700
CAS Number: 436799-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR