3-Bromo-5-(trifluoromethyl)pyridin-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-bromo-5-(trifluoromethyl)pyridin-2-ol, 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 3-bromo-5-(trifluoromethyl)pyridine-2-ol

A219972-1G A219972-25G
103818-828EA 11 USD
103818-828 103849-710 77088-292 77303-536
3-Bromo-5-(trifluoromethyl)pyridin-2-ol 98%
3-Bromo-5-(trifluoromethyl)pyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₃BrF₃NO
MW: 241.995 g/mol
Boiling Pt: 252.7 °C (760 mm Hg)
Melting Pt: 170…171 °C
Flash Pt: 106.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02691223
CAS Number: 76041-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR