3-Bromo-5-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A152031-5G A152031-1G A152031-25G
103804-340EA 5 USD
103804-340 103842-494 77299-922 77318-094 77318-096
3-Bromo-5-(trifluoromethyl)benzonitrile 97%
3-Bromo-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃N
Boiling Pt: 216 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09800708
CAS Number: 691877-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR