3-Bromo-5-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A142548-100G A142548-10G A142548-25G A142548-5G
103803-178EA 5 USD
103803-178 77190-496 77190-498 77299-302 77299-304 77316-404
3-Bromo-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
Boiling Pt: 284.3 °C (760 mm Hg)
Melting Pt: 132.3…132.8 °C
Flash Pt: 154.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03412186
CAS Number: 328-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR