3-Bromo-5-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed
Warning

A113621-100G A113621-10G A113621-25G A113621-1G
103798-256EA 5 USD
103798-256 103824-096 77189-186 77189-188 77189-190 77310-910
3-Bromo-5-(trifluoromethyl)benzaldehyde 98%
3-Bromo-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
Boiling Pt: 218.9 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08059506
CAS Number: 477535-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR