3-Bromo-4-pyridone 98%

Supplier: Ambeed
Warning

A157477-10G A157477-250MG A157477-1G A157477-5G
103805-010EA 10 USD
103805-010 103843-210 76973-378 76973-380 77319-034 77319-036
3-Bromo-4-pyridone 98%
3-Bromo-4-pyridone

Precaución: for Research Use Only

Formula: C₅H₄BrNO
Boiling Pt: 244.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01646107
CAS Number: 36953-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR