(3-Bromo-4-pyridyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-pyridinemethanol, 3-Bromo-4-(hydroxymethyl)pyridine

A208473-100MG A208473-10G A208473-250MG A208473-25G A208473-5G A208473-1G
103817-874EA 18 USD
103817-874 103848-560 77030-418 77030-420 77030-422 77086-876 77086-878 77303-070
(3-Bromo-4-pyridyl)methanol 95%
(3-Bromo-4-pyridyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.03 g/mol
Boiling Pt: 287.079 °C (760 mm Hg)
Melting Pt: 112…116 °C
Flash Pt: 127.421 °C
Storage Temperature: Ambient
MDL Number: MFCD11847285
CAS Number: 146679-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR