3-Bromo-4-nitropyridine 95%

Supplier: Ambeed
Warning

A123073-100MG A123073-250MG A123073-25G A123073-1G A123073-5G A123073-10G
103800-338EA 8 USD
103800-338 77108-674 77108-676 77108-678 77312-456 77312-458 77571-410
3-Bromo-4-nitropyridine 95%
3-Bromo-4-nitropyridine

Precaución: for Research Use Only

Formula: C5H3BrN2O2
MW: 202.99 g/mol
Boiling Pt: 235.7 °C (760 mm Hg)
Melting Pt: 62…66 °C
Flash Pt: 96.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03085767
CAS Number: 89364-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR