3-Bromo-4-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A311380-250MG A311380-1G
103814-662EA 72 USD
103814-662 103835-850 77114-624 77306-052
3-Bromo-4-nitrobenzaldehyde 97%
3-Bromo-4-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃
MW: 230.02 g/mol
Boiling Pt: 311.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00968939
CAS Number: 101682-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR