3-Bromo-4-methylthiophene 97%

Supplier: Ambeed
Warning

A506856-25G A506856-5G A506856-100G A506856-10G A506856-1G
103828-334EA 5 USD
103828-334 103839-436 76843-412 76843-414 77279-728 77279-730 77307-640
3-Bromo-4-methylthiophene 97%
3-Bromo-4-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅BrS
Boiling Pt: 181 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00130084
CAS Number: 30318-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR