3-Bromo-4-methylpyridin-2-ol 98%

Supplier: Ambeed
Warning

A185636-1G A185636-10G
103815-868EA 15 USD
103815-868 103846-438 77084-072 77260-360
3-Bromo-4-methylpyridin-2-ol 98%
3-Bromo-4-methylpyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839708
CAS Number: 18368-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR