3-Bromo-p-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-methylphenol, 1-Bromo-3-hydroxy-6-methylbenzene

A303828-10G A303828-1G A303828-5G A303828-25G A303828-100G
103814-394EA 5 USD
103814-394 103835-560 76787-806 76838-246 76838-248 77305-886 77668-348
3-Bromo-p-cresol 97%
3-Bromo-p-cresol

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.04
Boiling Pt: 244.6 °C (760 mm Hg)
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD08273793
CAS Number: 60710-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR