3-Bromo-p-toluonitrile 98%

Supplier: Ambeed
Warning

A223929-25G A223929-5G A223929-100G A223929-10G A223929-500G
103849-956EA 7 USD
103849-956 77088-584 77088-586 77275-560 77275-562 77275-564
3-Bromo-p-toluonitrile 98%
3-Bromo-p-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
Boiling Pt: 259 °C (760 mm Hg)
Melting Pt: 41…45 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD06797818
CAS Number: 42872-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR