3-Bromo-4-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A232868-100G A232868-10G A232868-250MG A232868-1G A232868-5G
103810-862EA 8 USD
103810-862 103850-618 77031-094 77031-096 77031-098 77089-560 77089-562
3-Bromo-4-methylbenzaldehyde 97%
3-Bromo-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Melting Pt: 47…52 °C
Flash Pt: 90.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04971197
CAS Number: 36276-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR