3-Bromo-p-toluidine 98%

Supplier: Ambeed
Warning

A296390-100G A296390-10G A296390-500G A296390-25G A296390-5G
103835-234EA 5 USD
103835-234 76772-522 76772-524 76772-526 76837-900 77305-690
3-Bromo-p-toluidine 98%
3-Bromo-p-toluidine

Precaución: for Research Use Only

Formula: C₇H₈BrN
Boiling Pt: 254…257 °C (760 mm Hg)
Melting Pt: 27…30 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134176
CAS Number: 7745-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR