3-Bromo-4-methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A213864-100MG A213864-250MG
103818-344EA 757 USD
103818-344 103849-056 76986-824 76986-826
3-Bromo-4-methyl-1H-pyrazole-5-carboxylic acid 95%
3-Bromo-4-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MW: 205.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22030963
CAS Number: 929554-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR