3-Bromo-4-methyl-1H-pyrazole 95%
Supplier: Ambeed
|
Warning
|
A239973-100MG
A239973-250MG
A239973-25G
A239973-1G
103811-322EA
42
USD
103811-322
103851-120
76976-418
76976-420
76976-422
77090-272
3-Bromo-4-methyl-1H-pyrazole 95%
3-Bromo-4-methyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₄H₅BrN₂ Boiling Pt: 271.6±20.0 °C (760 mm Hg) Melting Pt: 127…128 °C Flash Pt: 118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12755927 CAS Number: 5932-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...