3-Bromo-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A122215-100G A122215-10G A122215-25G A122215-1G A122215-5G
103800-190EA 5 USD
103800-190 77161-350 77161-352 77161-354 77312-330 77312-332
3-Bromo-4-methoxypyridine 98%
3-Bromo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Boiling Pt: 214.5 °C (760 mm Hg)
Flash Pt: 83.5 °C
Storage Temperature: Freezer
MDL Number: MFCD05664024
CAS Number: 82257-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR