3-Bromo-4-methoxyphenol 98%

Supplier: Ambeed
Warning

A118947-100G A118947-10G A118947-250MG A118947-1G A118947-5G A118947-25G
103799-140EA 15 USD
103799-140 77094-390 77094-392 77094-394 77297-650 77297-652 77592-316
3-Bromo-4-methoxyphenol 98%
3-Bromo-4-methoxyphenol

Precaución: for Research Use Only

Formula: C7H7BrO2
MW: 203.0300 g/mol
Boiling Pt: 281.1 °C (760 mm Hg)
Melting Pt: 47 °C
Flash Pt: 123.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11054143
CAS Number: 17332-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR