3-Bromo-p-anisaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-methoxybenzaldehyde

A115375-100G A115375-10G A115375-25G A115375-5G
103808-142EA 705 USD
103808-142 103824-412 77233-882 77233-884 77233-886 77311-228
3-Bromo-p-anisaldehyde 98%
3-Bromo-p-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 290.4 °C (760 mm Hg)
Melting Pt: 51…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00016599
CAS Number: 34841-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR