3-Bromo-p-anisidine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-methoxyaniline

A164473-100G A164473-10G A164473-500G A164473-5G A164473-1G A164473-25G
103805-866EA 9 USD
103805-866 103844-124 77273-468 77273-470 77273-472 77300-718 77320-292 77320-294
3-Bromo-p-anisidine 97%
3-Bromo-p-anisidine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
Boiling Pt: 282.2 °C (760 mm Hg)
Melting Pt: 62 °C
Flash Pt: 124.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04094339
CAS Number: 19056-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR