3-Bromo-4-iodobenzoic acid 97%

Supplier: Ambeed
Warning

A164277-100G A164277-10G A164277-250MG A164277-1G A164277-5G A164277-25G
103805-836EA 9 USD
103805-836 103844-096 76872-136 76872-138 76872-140 77300-700 77300-702 77320-254
3-Bromo-4-iodobenzoic acid 97%
3-Bromo-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.914 g/mol
Melting Pt: 242…243 °C
Storage Temperature: Ambient
MDL Number: MFCD06808825
CAS Number: 249647-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR