3-Bromo-4-hydroxybenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A121501-250MG
A121501-5G
A121501-1G
A121501-25G
A121501-500G
103800-082EA
6
USD
103800-082
76766-516
77297-838
77312-200
77571-276
77876-878
3-Bromo-4-hydroxybenzonitrile 98%
3-Bromo-4-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₄BrNO Boiling Pt: 271.1 °C (760 mm Hg) Melting Pt: 155…159 °C Flash Pt: 117.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143096 CAS Number: 2315-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...